Dissecting Pan and Vinyl Mat, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
63313
102091-090EA 67.8 USD
102091-090
Dissecting Pan and Vinyl Mat, Electron Microscopy Sciences
This dissecting pan is made from aluminum, which is rust free and easy to clean. It comes with a removable reusable vinyl pad, which accepts dissecting as well as push-pins to hold down your specimen during dissecting. The pad measures 131/8 x 93/8 x 21/4" thick.