Periodic acid 0,5% in aqueous solution

Supplier: Electron Microscopy Sciences
Warning

Synonyms: Orthoperiodic acidortho-Periodic acid

19324-05
101410-736EA 48.13 USD
101410-736
Periodic acid 0,5% in aqueous solution
Periodic acid
Formula: H₅IO₆
MW: 227.94 g/mol
MDL Number: MFCD00011344
CAS Number: 10450-60-9
https://us.vwr.com/stibo/web/std.lang.all/90/77/779077.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR