VIKEM® Vinyl Coated Lead Sheets, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64434-03 64434-05
100498-812EA 79.73 USD
100498-812 100498-816
VIKEM® Vinyl Coated Lead Sheets, Electron Microscopy Sciences
Weights Flask Weights
These soft lead sheets coated with Vikem vinyl have many uses for stabilizing laboratory equipment in water baths or any application where extra weight is needed. Small sheets are handy as a hold-down weight for note books, drawings or charts on the table top. Longer sheets may be bent into the shape of the glassware to keep them from sliding or tilting. Coated lead sheets are 2.2 mm (0.085") thick.