- Home
- Film Processing Supplies
- Saunders/LPL Enlargers, 4×5" Electron Microscopy Sciences
Saunders/LPL Enlargers, 4×5" Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
74871-10
74871-30
74870-20
74870-30
74870-10
74871-20
100494-116EA
4391.71
USD
100494-116
100494-114
100494-112
100494-110
100494-108
100494-106
Saunders/LPL Enlargers, 4×5" Electron Microscopy Sciences
Film Processing Supplies
Supplied with: Quartz-Halogen lamp
Regulated solid-state power supply. Swing away combination red safety filter and under-the-lens-holder. Does not include: A negative carrier, lens or lens mount (see accessories list). These enlargers have common a lamphouse and chassis configuration with different slide-in filtration modules, depending on the model. 4x5" enlargers accept all film formats from 35 mm to 4 x 5". The 4550XLG has a longer column and an extended chassis, a larger baseboard, and a higher output lamp.
Supplied with: Quartz-Halogen lamp. Regulated solid-state power supply. Swing away combination red safety filter and under-the-lens-holder. Does not include: A negative carrier, lens or lens mount (see accessories list).
MODEL 4550XLG ENLARGERS Additional features include being supplied with 250 watts, 82 volt EVW quartz-halogen lamp for 25% greater light output
Improved ventilation. A Whisper ToneTM muffling fan system for quiet operation. The fan is mounted on shock-dampening bearings to eliminate vibration. Extra-long column and oversized baseboard allows 20 x 24" enlargements of 35 mm negatives (using 50 mm lens). An extended carriage increases the distance from the column to the lens axis by 2 1/2" which allows for use of a larger easel (20 x 24").
Model 4550XLG Color Dichroic:
As described. Includes Dichroic Filtration Module with 0-200 yellow, 0-200 cyan, and 0-170 magenta stepless and continuously variable filtration
Permanent, fade-free dichroic filter. Color coded, illuminated filtration scale calibrated in 1cc increments
Memory-Lock™ while light focusing
Two f-stop light attenuator
Model 4550XLG Variable Contrast/Constant Exposure (VCCE):
As described. Includes a special Dichroic Filtration Modulecalibrated for the filtration requirements of Ilford Multigrade III variable contrast paper
Instantly set for the Ilford filter equivalent (conversion chart supplied for Kodak papers)
Automatic neutral density (ND) filter keeps exposure times constant throughout the contrast range
Model 4550XLG Black & White Diffusion:
As described with same quartz-halogen/diffusion system for fast B&W printing and the scratch and dust suppression characteristics of diffusion-type light sources
Model 4500-II Enlargers
Valuable 4x5" enlarger
Model 4500-II Color Dichroic
As described. Includes Dichroic Filtration Module with 0-200 yellow, 0-200 cyan, and 0-170 magenta stepless and continuously variable filtration
Supplied with 200watt/82volt (EYA) Quartz-Halogen lamp with integral dichroic reflector
Permanent, fade-free dichroic filters. Color-coded, illuminated filtration scales calibrated in 1cc increments
Memory-Locks(TM) instant while light focusing
Two f-stop light attenuator
Model 4500-II Contrast/Constant Exposure (VCCE)
Model 4500-II Black & White Diffusion MODEL 4550XLG ENLARGERS Additional features include being supplied with 250 watts, 82 volt EVW quartz-halogen lamp for 25% greater light output
Improved ventilation. A Whisper ToneTM muffling fan system for quiet operation. The fan is mounted on shock-dampening bearings to eliminate vibration. Extra-long column and oversized baseboard allows 20 x 24" enlargements of 35 mm negatives (using 50 mm lens). An extended carriage increases the distance from the column to the lens axis by 2 1/2" which allows for use of a larger easel (20 x 24").
Supplied with: Quartz-Halogen lamp. Regulated solid-state power supply. Swing away combination red safety filter and under-the-lens-holder. Does not include: A negative carrier, lens or lens mount (see accessories list).
MODEL 4550XLG ENLARGERS Additional features include being supplied with 250 watts, 82 volt EVW quartz-halogen lamp for 25% greater light output
Improved ventilation. A Whisper ToneTM muffling fan system for quiet operation. The fan is mounted on shock-dampening bearings to eliminate vibration. Extra-long column and oversized baseboard allows 20 x 24" enlargements of 35 mm negatives (using 50 mm lens). An extended carriage increases the distance from the column to the lens axis by 2 1/2" which allows for use of a larger easel (20 x 24").
Model 4550XLG Color Dichroic:
As described. Includes Dichroic Filtration Module with 0-200 yellow, 0-200 cyan, and 0-170 magenta stepless and continuously variable filtration
Permanent, fade-free dichroic filter. Color coded, illuminated filtration scale calibrated in 1cc increments
Memory-Lock™ while light focusing
Two f-stop light attenuator
Model 4550XLG Variable Contrast/Constant Exposure (VCCE):
As described. Includes a special Dichroic Filtration Modulecalibrated for the filtration requirements of Ilford Multigrade III variable contrast paper
Instantly set for the Ilford filter equivalent (conversion chart supplied for Kodak papers)
Automatic neutral density (ND) filter keeps exposure times constant throughout the contrast range
Model 4550XLG Black & White Diffusion:
As described with same quartz-halogen/diffusion system for fast B&W printing and the scratch and dust suppression characteristics of diffusion-type light sources
Model 4500-II Enlargers
Valuable 4x5" enlarger
Model 4500-II Color Dichroic
As described. Includes Dichroic Filtration Module with 0-200 yellow, 0-200 cyan, and 0-170 magenta stepless and continuously variable filtration
Supplied with 200watt/82volt (EYA) Quartz-Halogen lamp with integral dichroic reflector
Permanent, fade-free dichroic filters. Color-coded, illuminated filtration scales calibrated in 1cc increments
Memory-Locks(TM) instant while light focusing
Two f-stop light attenuator
Model 4500-II Contrast/Constant Exposure (VCCE)
Model 4500-II Black & White Diffusion MODEL 4550XLG ENLARGERS Additional features include being supplied with 250 watts, 82 volt EVW quartz-halogen lamp for 25% greater light output
Improved ventilation. A Whisper ToneTM muffling fan system for quiet operation. The fan is mounted on shock-dampening bearings to eliminate vibration. Extra-long column and oversized baseboard allows 20 x 24" enlargements of 35 mm negatives (using 50 mm lens). An extended carriage increases the distance from the column to the lens axis by 2 1/2" which allows for use of a larger easel (20 x 24").
Model 4550XLG Color Dichroic:
As described. Includes Dichroic Filtration Module with 0-200 yellow, 0-200 cyan, and 0-170 magenta stepless and continuously variable filtration
Permanent, fade-free dichroic filter. Color coded, illuminated filtration scale calibrated in 1cc increments
Memory-Lock™ while light focusing
Two f-stop light attenuator
As described. Includes Dichroic Filtration Module with 0-200 yellow, 0-200 cyan, and 0-170 magenta stepless and continuously variable filtration
Permanent, fade-free dichroic filter. Color coded, illuminated filtration scale calibrated in 1cc increments
Memory-Lock™ while light focusing
Two f-stop light attenuator
Model 4550XLG Variable Contrast/Constant Exposure (VCCE):
As described. Includes a special Dichroic Filtration Modulecalibrated for the filtration requirements of Ilford Multigrade III variable contrast paper
Instantly set for the Ilford filter equivalent (conversion chart supplied for Kodak papers)
Automatic neutral density (ND) filter keeps exposure times constant throughout the contrast range
As described. Includes a special Dichroic Filtration Modulecalibrated for the filtration requirements of Ilford Multigrade III variable contrast paper
Instantly set for the Ilford filter equivalent (conversion chart supplied for Kodak papers)
Automatic neutral density (ND) filter keeps exposure times constant throughout the contrast range
Model 4550XLG Black & White Diffusion:
As described with same quartz-halogen/diffusion system for fast B&W printing and the scratch and dust suppression characteristics of diffusion-type light sources
As described with same quartz-halogen/diffusion system for fast B&W printing and the scratch and dust suppression characteristics of diffusion-type light sources
Model 4500-II Enlargers
Valuable 4x5" enlarger
Model 4500-II Color Dichroic
As described. Includes Dichroic Filtration Module with 0-200 yellow, 0-200 cyan, and 0-170 magenta stepless and continuously variable filtration
Supplied with 200watt/82volt (EYA) Quartz-Halogen lamp with integral dichroic reflector
Permanent, fade-free dichroic filters. Color-coded, illuminated filtration scales calibrated in 1cc increments
Memory-Locks(TM) instant while light focusing
Two f-stop light attenuator
Model 4500-II Contrast/Constant Exposure (VCCE)
Model 4500-II Black & White Diffusion
Valuable 4x5" enlarger
Model 4500-II Color Dichroic
As described. Includes Dichroic Filtration Module with 0-200 yellow, 0-200 cyan, and 0-170 magenta stepless and continuously variable filtration
Supplied with 200watt/82volt (EYA) Quartz-Halogen lamp with integral dichroic reflector
Permanent, fade-free dichroic filters. Color-coded, illuminated filtration scales calibrated in 1cc increments
Memory-Locks(TM) instant while light focusing
Two f-stop light attenuator
Model 4500-II Contrast/Constant Exposure (VCCE)
Model 4500-II Black & White Diffusion
New Product
Limited Time Sale
Select items below are currently on sale
Enter Certificate Details
Inquire for Price
Learn more
About VWR
Item(s) Added to Cart