Components for Series 4 × 5 Enlargers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
74872-10 74873-22 74873-24 74873-26 74873-23 74872-30 74872-20
100494-138EA 245.18 USD
100494-138 100494-122 100494-134 100494-120 100494-132 100494-130 100494-118
Components for Series 4 × 5 Enlargers, Electron Microscopy Sciences
Film Processing Supplies
Interchangeable Slide-in Filtration Modules that fits all Saunders/LPL 4550XLG and 4500-II enlargers
  • All aluminum precision carriers
  • Hinged sandwich design for fast handling and loading
  • Guide pins automatically position negatives in line with carrier aperture
  • Offset tabs permits rapid opening of carrier
  • All 4500 Carriers can be used on Omega D5 Enlarger
Both glass and glassless are available as follows:
Interchangeable Slide-in Filtration Modules that fits all Saunders/LPL 4550XLG and 4500-II enlargers. Both models are easily converted by switching the modules.
Negative Carriers For Both 4550XLG and 4500-II
Negative Carriers For Both 4550XLG and 4500-II
Both models are easily converted by switching the modules.