Dumont Tweezers Style 5/45 SA ESD, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
0304-5/45SAESD-CO
102091-694EA 57.31 USD
102091-694
Dumont Tweezers Style 5/45 SA ESD, Electron Microscopy Sciences
Forceps
Thin, 109 mm tips, bent 45°.
Antimagnetic is composed of C, Cr, Ni and Mo. It offers a good resistance to corrosion thanks to its high concentration of molybdenum and is 80% antimagnetic but is not as hard as Inox. Antimagnetic is resistant up to temperatures of around 400°C (DIN 50 914) and is suitable for autoclave sterilization at 270°C.