Would you like to log in again?
Avantor is taking action. Our response to the Novel Coronavirus outbreak.
Tweezers Protection Cap, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72904-12
100501-910PK 2.34 USD InStock
100501-910
Tweezers Protection Cap, Electron Microscopy Sciences
Forceps
These caps which are in the shape of the tweezer tip are made from Polyethylene and are used to protect the tweezers tips from damage.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty