Dumont Diamond Tips Straight Tweezers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
78100
100502-140EA 67.39 USD
100502-140
Dumont Diamond Tips Straight Tweezers, Electron Microscopy Sciences
Forceps
Fine Tip :Tip measures 0.25 by 0.25 mm.
Dumont Stainless Steel tweezers coated with an extremely fine diamond dust in the inside tip. They are essential in applications where a tight grip is necessary. They are also available with an improved locking system. They are 150 mm (5 1/2") long. They may not be suitable for use with EM grids.
Fine Tip :Tip measures 0.25 by 0.25 mm.
Medium Tip: Tip measures 0.5 by 0.5 mm.