Would you like to log in again?
Air Valves with Filter Function for Safety Caps, S.C.A.T.
Supplier: S.C.A.T
197010 197011 112010 112110 117010 117011
10040-110PK 442.52 USD InStock
10040-110 10060-300 10040-150 10060-298 10040-108 10040-248
Air Valves with Filter Function for Safety Caps, S.C.A.T.
Closures Safety Caps and Safety Waste Caps
All Safety Caps are equipped with an air valve, combining air valve and filter function into one unit
Because it absorbs dust and contaminant particles from the surrounding inflowing air, the air valve must be replaced every six months to guarantee flawless operation and full protection. All options are UNF 1/4".
The air valve provides trouble-free ventilation during fluid extraction and keeps hazardous gases inside the reservoir. This protects the solvent from contamination.
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty