SP Bel-Art Belpen™ Markers, Bel-Art Products, a part of SP
Supplier: Bel-Art
F13374-0002 F13374-0000
13196-060PK 35.23 USD
13196-060 13196-064
SP Bel-Art Belpen™ Markers, Bel-Art Products, a part of SP
Pens Ink Pens
Felt tip marking pens are perfect for labeling all types of glassware and plastics.
  • Felt tip design
  • Permanent, quick-drying smudge-proof ink
  • Dried ink is waterproof and autoclavable
  • With handy pocket clip
Pen is 13.3 cm (5¹/₄") long.