CO2 Incubators, C170, BINDER
9040-0115 9040-0116 9040-0113 9040-0112
75783-584EA 8927.49 USD
75783-584 75783-586 75783-580 75783-582
CO2 Incubators, C170, BINDER
CO2 incubators provide optimal growth conditions and humidity without condensation for cell cultures.
  • Tightly sealed inner tempered safety glass
  • Exterior optional lockable door handle
Overnight, the automatic hot air sterilization cycle operates at 180°C removing contamination
Models feature a drift-free IR sensor for absolute CO2 management and LCD microprocessor. It also includes an automatic diagnostic system with visual and audible alarms and a zero voltage alarm contact. The unit also has an independent safety device class 3.1 with optical and audible temperature alarms.
The seamless deep-drawn stainless steel inner chamber makes cleaning fast and easy. It features a removable shelf support system preventing accumulation of particulate matter.
Certifications: The 120V units are UL listed.
Ordering information: The incubators are available with a base platform stand and rubber pads for holding one incubator 8"" off the ground. The stand is available with or without castors. Vibration-free stacking frames are also available for the direct and safe stacking of two CO2 incubators. Frames are designed on castors with stop brake and wedge equipment.