Would you like to log in again?
KBW Series Growth Chambers, BINDER
Supplier: BINDER INC
9020-0256 9020-0180
80079-056EA 21935.3 USD InStock
80079-056 89182-866
KBW Series Growth Chambers, BINDER
Testing Chambers Plant Growth Chambers
Electronically Controlled APT.line preheating chamber and refrigerating system assuring temperature accuracy and reproducible results
Inner glass door with sealing. MP Controller with 2 programs with 10 sections each, alternatively switchable to 1 program with 20 sections
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty