Would you like to log in again?
Wig-L-Bug Grinding Mill, International Crystal Labs
0013-5236
100213-258EA 1736.31 USD InStock
100213-258
Wig-L-Bug Grinding Mill, International Crystal Labs
Mills
The Wig-L-Bug is an invaluable accessory for the spectroscopy laboratory
It is ideal for mixing and pulverizing samples for use in a KBr matrix that will be pressed into a pellet and for mulling techniques. Sample preparation is reproducible from sample to sample by replicating the cycle time, which is controlled by a 30 second timer. Most samples can be prepared in 10 seconds.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty