Anti-CD20 Mouse Monoclonal Antibody [clone: L26]

Supplier: Diagnostic BioSystems

MOB004-05 PDM004 MOB004
95024-322EA 278.23 USD
95024-322 99990-822 99990-170
Anti-CD20 Mouse Monoclonal Antibody [clone: L26]
Antibodies
This antibody is specific to a 33 kDa polypeptide present on the majority of B cells in peripheral blood and lymphoid tissue. No reactivity with other hematopoietic cells has been observed.

Type:
Antigen: CD20
Clonality: Monoclonal
Clone: L26
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR