Anti-CD15 Mouse Monoclonal Antibody [clone: MMA]

Supplier: Diagnostic BioSystems

PDM127 MOB365 MOB365-05
99990-682EA 489.38 USD
99990-682 99991-026 95024-316
Anti-CD15 Mouse Monoclonal Antibody [clone: MMA]
Antibodies
This antibody reacts with a 220 kDa protein. CD15 plays a role in mediating phagocytosis, bactericidal activity and chemotaxis. It is present on >95% of granulocytes including neutrophils and eosinophils and to a lesser degree on monocytes. CD15 is also expressed in Reed-Sternberg cells and some epithelial cells. CD15 is very useful in the identification of Hodgkin disease.

Type:
Antigen: CD15
Clonality: Monoclonal
Clone: MMA
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR