Anti-CD117/C-Kit Rabbit Polyclonal Antibody

Supplier: Diagnostic BioSystems

RP063-05 RP063 PDR045
99991-302EA 478.2 USD
99991-302 95024-954 99991-120
Anti-CD117/C-Kit Rabbit Polyclonal Antibody
Antibodies
This antibody reacts with a 145 kDa protein. CD117/c-kit is a transmembrane receptor tyrosine kinase and is expressed in many tissues and cells. CD117/c-kit is involved in the development of several lineages of stem cells, such as germ cells, neural crest derived melanocytes and hematopoietic precursor cells. This antibody can be used in the identification of gastrointestinal stromal tumors.

Type:
Antigen: CD117/C-Kit
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR