Anti-CH Mouse Monoclonal Antibody [clone: CALP]

Supplier: Diagnostic BioSystems

MOB345 MOB345-05
95024-266EA 570.12 USD
95024-266 99990-646
Anti-CH Mouse Monoclonal Antibody [clone: CALP]
Antibodies
This antibody reacts with a 34 kDa protein known as calponin. Calponin, a calmodulin, binds tropomyosin and F-actin and is thought to be involved in the regulation of smooth muscle contraction. Expression of calponin is restricted to smooth muscle cells. Two isoforms of calponin exist whose molecular weights are 34 KD and 29 kDa. 29 kDa isoform is primarily restricted to the muscle of the urogenital tract.

Type:
Antigen: CH
Clonality: Monoclonal
Clone: CALP
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR