Anti-CALCA 0 Polyclonal Antibody

Supplier: Diagnostic BioSystems

RP050 RP050-05 PDR024
99991-288EA 540.3 USD
99991-288 95024-920 99991-088
Anti-CALCA 0 Polyclonal Antibody
Antibodies
This antibody stains C cells of thyroid.

Type:
Antigen: CALCA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: 0
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR