Super HT PAP Pens, Biotium
Supplier: Biotium
22005 22006
89427-056EA 175.27 USD
89427-056 89427-058
Super HT PAP Pens, Biotium
Stains Microscopy and staining reagents
The Super HT PAP pen draws a water repellent circle around slide mounted tissue and prevents the waste of valuable reagents by keeping the liquid pooled in a single droplet
The regular sized Super HT PAP Pen can be used in about 800 applications, while the mini sized Super HT PAP Pen will last about 400 applications.