Would you like to log in again?
Avantor is taking action. Our response to the Novel Coronavirus outbreak.
Stripettor™ Plus Pipette Controller, Corning®
4090
89133-506CS 484.45 USD InStock
89133-506
Stripettor™ Plus Pipette Controller, Corning®
Pipets Pipet Controllers
The Corning® Stripettor™ Plus is comfortable, lightweight, and designed for use with most brands of serological pipettes
Speed range is adjustable via a control wheel on the back of the instrument, and aspirating and dispensing can be finely controlled by the pressure-sensitive buttons. When fully charged, the high capacity Lithium-Ion battery allows continuous pipetting for up to 4.5 hours.
Ordering information: A convenient wall hanger, charging stand, and spare hydrophobic PTFE 0.2 µm filters are included in purchase of the Stripiettor™ Plus.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty