Would you like to log in again?
Corning® 96- and 384-Well High Content Imaging Microplates
Supplier: Corning
Corning®
4681 4680
10037-554CS 695.72 USD InStock
10037-554 10037-552
Corning® 96- and 384-Well High Content Imaging Microplates
Microplates Cell Culture Plates
With an ultra-clear film, a 127μm film thickness, and an unprecedented flatness (whole plate and intra-well), these microplates are ideal for high resolution cellular imaging applications
Both 96-well and 384-well High Content Imaging Mircoplates are black with a clear bottom. Plates are supplied with lids.
The microplate and film are manufactured from cyclic olefin copolymer, which offers excellent optical properties, chemical resistance, and mechanical stability.
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty