Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Hydroxypyrimidine hydrochloride for synthesis

Supplier: MilliporeSigma

Synonyms: 2-Pyrimidinol Hydrochloride2(1H)-pyrimidinone monohydrochloride

8.43938.0100 8.43938.0100 8.43938.0025 8.43938.0025
EM8.43938.0100CS 18819.31 USD InStock
CA8.43938.0100 EM8.43938.0100 CA8.43938.0025 EM8.43938.0025
2-Hydroxypyrimidine hydrochloride for synthesis
2-Hydroxypyrimidine hydrochloride


Formula: C₄H₄N₂O
MW: 132,55 g/mol
MDL Number: MFCD00012781
CAS Number: 38353-09-2
Order Now
Specification Test Results
Solubility (20 °C) soluble
Melting Point 200 - 205 °C
Molar Mass 132.55 g/mol


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty