Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.

1-(4-Fluorophenyl)piperazine dihydrochloride for synthesis

Supplier: MilliporeSigma


8.43931.0005 8.43931.0005 8.43931.0025 8.43931.0025
EM8.43931.0005CS 34100.81 USD InStock
CA8.43931.0005 EM8.43931.0005 CA8.43931.0025 EM8.43931.0025
1-(4-Fluorophenyl)piperazine dihydrochloride for synthesis
1-(4-Fluorophenyl)piperazine dihydrochloride


Formula: C₁₀H₁₅Cl₂FN₂
MW: 253,15
MDL Number: MFCD00012765
CAS Number: 64090-19-3
EINECS: 000-000-0
Order Now
Specification Test Results
Melting Point 233 - 237 °C
Molar Mass 253.14 g/mol


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty