Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.

Triisooctylamine for synthesis

Supplier: MilliporeSigma

Warning

8.43805.0500 8.43805.0100 8.43805.0500 8.43805.0100
EM8.43805.0100CS 1146.21 USD InStock
CA8.43805.0500 EM8.43805.0100 EM8.43805.0500 CA8.43805.0100
Triisooctylamine for synthesis
Triisooctylamine


Formula: C₂₄H₅₁N
MW: 353.68 g/mol
Density: 0.8 g/cm³ (20 °C)
CAS Number: 25549-16-0
EINECS: 247-092-0
UN: 3082
ADR: 9,III
Order Now
Specification Test Results
Solubility  - 1 g/l (20 °C)
Molar Mass 353.68 g/mol
Boiling Point 204 °C
Flash Point 110 °C DIN 51758
Density 0.800 g/cm3 (20 °C)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty