Trimellitic anhydride chloride for synthesis, Sigma-Aldrich®

Danger

Synonyms: 1,2,4-Benzenetricarboxylic acid 1,2-anhydride 4-chloride1,3-Dioxo-1,3-dihydroisobenzofuran-5-carbonyl chloride

M-Clarity™ quality level = MQ200

8.43764.0025 8.43764.0100 8.43764.0100 8.43764.0025
EM8.43764.0025CS 5612.76 USD
CA8.43764.0100 CA8.43764.0025 EM8.43764.0025 EM8.43764.0100
Trimellitic anhydride chloride for synthesis, Sigma-Aldrich®
Trimellitic anhydride chloride
Formula: C₉H₃ClO₄
MW: 210.57 g/mol
MDL Number: MFCD00005924
CAS Number: 1204-28-0
EINECS: 214-874-8
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Solubility (20 °C) Hydrolysis
Melting Point 65 - 68 °C
Molar Mass 210.57 g/mol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR