Sodium triacetoxyborohydride for synthesis

Supplier: MilliporeSigma

8.43705.0025 8.43705.0025 8.43705.0100 8.43705.0100
EM8.43705.0025CS 2030.32 USD InStock
CA8.43705.0025 EM8.43705.0025 EM8.43705.0100 CA8.43705.0100
Sodium triacetoxyborohydride for synthesis
Sodium triacetoxyborohydride
Formula: C₆H₁₀BNaO₆
MW: 211.94 g/mol
Melting Pt: 114…118 °C
MDL Number: MFCD00012211
CAS Number: 56553-60-7
EINECS: 000-000-0
UN: 3396
ADR: 4.3,II

Order Now

Specification Test Results

Melting Point 110 - 114 °C
Molar Mass 211.94 g/mol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR