1-Ethyl-2-pyrrolidone for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 1-Ethyl-2-pyrrolidinoneN-Ethyl-2-pyrrolidone1-Ethylpyrrolidin-2-one

M-Clarity™ quality level = MQ200

8.42196.0025 8.42196.0025
EM8.42196.0025CS 5207 USD
EM8.42196.0025 CA8.42196.0025
1-Ethyl-2-pyrrolidone for synthesis, Sigma-Aldrich®
1-Ethyl-2-pyrrolidone
Formula: C₆H₁₁NO
MW: 113.16 g/mol
Boiling Pt: 97 °C (1013 hPa)
Density: 0.9979 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003199
CAS Number: 2687-91-4
EINECS: 220-250-6

Order Now

Specification Test Results

Solubility (20 °C) soluble
Molar Mass 113.16 g/mol
Boiling Point 97 °C (33 hPa)
Vapor Pressure 0.5 hPa (32 °C)
Flash Point 91 °C
Explosion Limit 1.3 - 7.7 %(V)
Density 1.0 g/cm3 (20 °C)
pH 8.2 (100 g/l, H2O, 23 °C)
Ignition Point 250 °C DIN 51794
Viscosity Kinematic 2.1 mm2/s (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR