Acetylcholine iodide for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
(2-Acetoxyethyl)trimethylammonium iodide
, 2-Acetoxy-N,N,N-trimethylethan-1-aminium iodide
M-Clarity™ quality level = MQ200
8.41870.0025
8.41870.0025
EM8.41870.0025CS
22918
USD
EM8.41870.0025
CA8.41870.0025
Acetylcholine iodide for synthesis, Sigma-Aldrich®
Acetylcholine iodide
Formula:
H₃CC(O)O(CH₂)₂N(CH₃)₃I MW: 273.11 g/mol Melting Pt: 163…165 °C Density: 1.67 g/cm³ (20 °C) Storage Temperature: Freezer |
MDL Number:
MFCD00011815 CAS Number: 2260-50-6 EINECS: 218-862-3 |
Specification Test Results
Melting Point | 162 - 165 °C |
Molar Mass | 273.11 g/mol |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...