2-Thienylacetyl chloride for synthesis, Sigma-Aldrich®

Danger

Synonyms: Thiophene-2-acetyl chloride2-Thiopheneacetyl chloride

M-Clarity™ quality level = MQ200

8.41865.0025 8.41865.0025
EM8.41865.0025CS 33037.83 USD
CA8.41865.0025 EM8.41865.0025
2-Thienylacetyl chloride for synthesis, Sigma-Aldrich®
2-Thienylacetyl chloride
Formula: C₆H₅ClOS
MW: 160.62
MDL Number: MFCD00005456
CAS Number: 39098-97-0
EINECS: 254-290-0
UN: 1760
ADR: 8,III

Order Now

Specification Test Results

Molar Mass 160.62 g/mol
Boiling Point 130 - 135 °C (120 hPa)
Flash Point 101 °C
Density 1.303 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR