3,5-Dichloroaniline for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.41790.0100 8.41790.0100
EM8.41790.0100CS 1936 USD
CA8.41790.0100 EM8.41790.0100
3,5-Dichloroaniline for synthesis, Sigma-Aldrich®
3,5-Dichloroaniline
Formula: C₆H₅Cl₂N
MW: 162.02 g/mol
Boiling Pt: 262 °C (1013 hPa)
Melting Pt: 51 °C
Density: 1.58 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007774
CAS Number: 626-43-7
EINECS: 210-948-9
UN: 3442
ADR: 6.1,II

Order Now

Specification Test Results

Solubility 0.78 g/l
Melting Point 46 - 53 °C
Molar Mass 162.02 g/mol
Boiling Point 259 - 260 °C (1013 hPa)
Vapor Pressure 40 hPa (150 °C)
Flash Point 133 °C
Density 1.58 g/cm3 (20 °C)
Ignition Point 620 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR