Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,6-Dibromophenol for synthesis

Warning
Synonyms: 1,3-Dibromo-2-hydroxybenzene

8.41765.0001 8.41765.0001
EM8.41765.0001CS 3287.73 USD InStock
CA8.41765.0001 EM8.41765.0001
2,6-Dibromophenol for synthesis
2,6-Dibromophenol


Formula: C₆H₄Br₂O
MW: 251.91 g/mol
Boiling Pt: 255 °C (1013 hPa)
Melting Pt: 55 - 56 °C
Storage Temperature: Ambient
MDL Number: MFCD00002152
CAS Number: 608-33-3
EINECS: 210-161-0
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Melting Point 55 - 57 °C
Molar Mass 251.9 g/mol
Boiling Point 255 - 256 °C (987 hPa)

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty