3-Phenylpyridine for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
M-Clarity™ quality level = MQ200
8.41759.0005
8.41759.0005
EM8.41759.0005CS
9245
USD
EM8.41759.0005
CA8.41759.0005
3-Phenylpyridine for synthesis, Sigma-Aldrich®
3-Phenylpyridine
Formula:
C₁₁H₉N MW: 155.2 g/mol Boiling Pt: 273…274 °C (1013 hPa) Melting Pt: 76…77 °C Density: 1.08 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006380 CAS Number: 1008-88-4 EINECS: 213-762-6 |
Specification Test Results
Assay (GC, area%) | ≥ 97.0 % (a/a) |
Density (d 20 °C/ 4 °C) | 1.090 - 1.094 |
Identity (IR) | passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...