Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromobutyric acid for synthesis

Danger
Synonyms: 4-Bromobutanoic acid

8.41654.0025 8.41654.0025
EM8.41654.0025CS 14566.5 USD InStock
CA8.41654.0025 EM8.41654.0025
4-Bromobutyric acid for synthesis
4-Bromobutyric acid


Formula: BrH₂C(CH₂)₂COOH
MW: 167 g/mol
Boiling Pt: 249 °C (1013 hPa)
Melting Pt: 29 °C
MDL Number: MFCD00002817
CAS Number: 2623-87-2
EINECS: 220-083-9
UN: 3261
ADR: 8,III
Order Now
Specification Test Results
Melting Point 29 °C
Molar Mass 167 g/mol
Boiling Point 129 - 131 °C (14.7 hPa)
Flash Point 115 °C

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty