Tricarballylic acid for synthesis

Supplier: MilliporeSigma

Warning

Synonyms: Propane-1,2,3-tricarboxylic acid1,2,3-Propantricarboxylic acid

8.41650.0010 8.41650.0010
EM8.41650.0010CS 71756.56 USD InStock
CA8.41650.0010 EM8.41650.0010
Tricarballylic acid for synthesis
Tricarballylic acid
Formula: C₆H₈O₆
MW: 176.13 g/mol
Boiling Pt: 266 °C (1013 hPa)
Melting Pt: 160 °C
Storage Temperature: Ambient
MDL Number: MFCD00002723
CAS Number: 99-14-9
EINECS: 202-733-3
Merck Index: 12,09752

Order Now

Specification Test Results

Solubility 500 g/l (18 °C)
Melting Point 160 °C
Molar Mass 176.12 g/mol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR