4-Bromo-1-butene for synthesis

Supplier: MilliporeSigma

8.41642.0005 8.41642.0005
EM8.41642.0005CS 10843.03 USD InStock
CA8.41642.0005 EM8.41642.0005
4-Bromo-1-butene for synthesis
4-Bromo-1-butene
Formula: C₄H₇Br
MW: 135 g/mol
Boiling Pt: 98 °C (1013 hPa)
Density: 1.33 g/cm³ (20 °C)
Flash Pt: 9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000258
CAS Number: 5162-44-7
EINECS: 225-937-4
UN: 1992
ADR: 3,II

Order Now

Specification Test Results

Solubility 0.76 g/l (25 °C)
Molar Mass 135 g/mol
Boiling Point 98 - 100 °C
Flash Point 1 °C
Density 1.33 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR