Dipropyl sulfide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.41603.0025 8.41603.0025
EM8.41603.0025CS 8055 USD
CA8.41603.0025 EM8.41603.0025
Dipropyl sulfide for synthesis, Sigma-Aldrich®
Dipropyl sulphide
Formula: C₆H₁₄S
MW: 118.24 g/mol
Boiling Pt: 146 °C (1013 hPa)
Melting Pt: –102 °C
Density: 0.83 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009379
CAS Number: 111-47-7
EINECS: 203-873-8
UN: 1993
ADR: 3,II
Merck Index: 12,08053

Order Now

Specification Test Results

Melting Point -102 °C
Molar Mass 118.23 g/mol
Boiling Point 141 - 142 °C
Vapor Pressure 8.6 hPa
Flash Point 28 °C
Explosion Limit 1 - 51 %(V)
Density 0.838 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR