2-Sulfobenzoic cyclic anhydride for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 2-sulphobenzoic acid cyclic anhydride2-sulphobenzoic anhydride

M-Clarity™ quality level = MQ200

8.41501.0025 8.41501.0025
EM8.41501.0025CS 8290 USD
CA8.41501.0025 EM8.41501.0025
2-Sulfobenzoic cyclic anhydride for synthesis, Sigma-Aldrich®
2-Sulphobenzoic cyclic anhydride
Formula: C₇H₄O₄S
MDL Number: MFCD00005879
CAS Number: 81-08-3
EINECS: 201-322-6
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Solubility (20 °C) (decomposition)
Melting Point 116 - 122 °C
Molar Mass 184.17 g/mol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR