Propargyltriphenylphosphonium bromide for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
Triphenyl(prop-2-yn-1-yl)phosphonium bromide
, 2-Propinyl-triphenylphosphonium bromide
M-Clarity™ quality level = MQ200
8.41338.0005
8.41338.0005
EM8.41338.0005CS
138818
USD
EM8.41338.0005
CA8.41338.0005
Propargyltriphenylphosphonium bromide for synthesis, Sigma-Aldrich®
Propargyltriphenylphosphonium bromide
Formula:
C₂₁H₁₈BrP MW: 381.26 g/mol |
MDL Number:
MFCD00011811 CAS Number: 2091-46-5 EINECS: 218-242-2 UN: 3464 ADR: 6.1,III |
Specification Test Results
Melting Point | 177 - 178 °C |
Molar Mass | 381.25 g/mol |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...