1-Pyrenebutyric acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.41269.0001 8.41269.0001
EM8.41269.0001CS 78743 USD
EM8.41269.0001 CA8.41269.0001
1-Pyrenebutyric acid for synthesis, Sigma-Aldrich®
1-Pyrenebutyric acid
Formula: C₂₀H₁₆O₂
MW: 288.35 g/mol
MDL Number: MFCD00004141
CAS Number: 3443-45-6
EINECS: 222-354-7

Order Now

Specification Test Results

Solubility (20 °C) practically insoluble
Melting Point 185 - 189 °C
Molar Mass 288.34 g/mol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR