Methyl cyclopropanecarboxylate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Methylcyclopropanecarboxylate

M-Clarity™ quality level = MQ200

8.41225.0025 8.41225.0025
EM8.41225.0025CS 6325 USD
CA8.41225.0025 EM8.41225.0025
Methyl cyclopropanecarboxylate for synthesis, Sigma-Aldrich®
Methyl cyclopropanecarboxylate
Formula: C₅H₈O₂
MW: 100.12 g/mol
MDL Number: MFCD00001280
CAS Number: 2868-37-3
EINECS: 220-690-9
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Molar Mass 100.12 g/mol
Boiling Point 118 - 119 °C
Flash Point 17 °C
Density 0.983 g/cm3 (20 °C)
Ignition Point 435 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR