Desyl chloride for synthesis, Sigma-Aldrich®

Danger

Synonyms: 2-Chloro-2-phenylacetophenone

M-Clarity™ quality level = MQ200

8.41176.0025 8.41176.0025
EM8.41176.0025CS 21498.28 USD
EM8.41176.0025 CA8.41176.0025
Desyl chloride for synthesis, Sigma-Aldrich®
Desyl chloride
Formula: C₁₄H₁₁ClO
MDL Number: MFCD00000858
CAS Number: 447-31-4
EINECS: 207-181-7
UN: 1759
ADR: 8,II

Order Now

Specification Test Results

Melting Point 67 - 68 °C
Molar Mass 230.69 g/mol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR