Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.

2,6-Diisopropylaniline for synthesis


8.41095.0100 8.41095.0500 8.41095.0500 8.41095.0100
EM8.41095.0100CS 474.88 USD InStock
EM8.41095.0100 CA8.41095.0500 EM8.41095.0500 CA8.41095.0100
2,6-Diisopropylaniline for synthesis
2,6-Diisopropylaniline


Formula: C₁₂H₁₉N
MW: 177.29 g/mol
Boiling Pt: 257 °C (1013 hPa)
Melting Pt: -45 °C
Density: 0,94 g/cm³ (20 °C)
Storage Temperature: Ambient
CAS Number: 24544-04-5
EINECS: 246-305-4
Order Now
Specification Test Results
Solubility  - 0.2 g/l (20 °C)
Melting Point -45 °C
Molar Mass 177.29 g/mol
Boiling Point 257 °C (1013 hPa)
Flash Point 125 °C
Density 0.94 g/cm3 (20 °C)
Ignition Point 400 °C DIN 51794

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty