2-Isopropylphenol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.40160.0100 8.40160.0100
EM8.40160.0100CS 784 USD
CA8.40160.0100 EM8.40160.0100
2-Isopropylphenol for synthesis, Sigma-Aldrich®
2-Isopropylphenol
Formula: C₉H₁₂O
MW: 136.19 g/mol
Boiling Pt: 213 °C (1013 hPa)
Melting Pt: 14…16 °C
Density: 0.995 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002224
CAS Number: 88-69-7
EINECS: 201-852-8
UN: 3145
ADR: 8,II

Order Now

Specification Test Results

Solubility 0.22 g/l (20 °C)
Melting Point 14 - 16 °C
Molar Mass 136.19 g/mol
Boiling Point 212 - 214 °C (1013 hPa)
Vapor Pressure  - 0.066 hPa (25 °C)
Flash Point 88 °C
Density 1.01 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR