2-Ethoxyethyl methacrylate stabilized for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
Methacrylic acid 2-Ethoxyethyl ester
, Ethylene glycol monoethyl ether Methacrylate
M-Clarity™ quality level = MQ200
8.40085.0025
8.40085.0025
EM8.40085.0025CS
4824
USD
EM8.40085.0025
CA8.40085.0025
2-Ethoxyethyl methacrylate stabilized for synthesis, Sigma-Aldrich®
2-Ethoxyethyl methacrylate
Formula:
C₈H₁₄O₃ MW: 158.1950 g/mol |
MDL Number:
MFCD00009249 CAS Number: 2370-63-0 EINECS: 219-135-3 |
Specification Test Results
Solubility | 13 g/l (20 °C) |
Molar Mass | 158.19 g/mol |
Boiling Point | 205 °C |
Vapor Pressure | 1.5 hPa (20 °C) |
Flash Point | 74 °C |
Density | 0.96 g/cm3 (20 °C) |
Ignition Point | 209 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...