Tetraoctylphosphonium bromide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Tetra-n-octylphosphonium bromide

M-Clarity™ quality level = MQ200

8.40068.0010 8.40068.0010
EM8.40068.0010CS 6868.45 USD
CA8.40068.0010 EM8.40068.0010
Tetraoctylphosphonium bromide for synthesis, Sigma-Aldrich®
Tetraoctylphosphonium bromide
Formula: C₃₂H₆₈BrP
MW: 563.77 g/mol
MDL Number: MFCD00210101
CAS Number: 23906-97-0
EINECS: 245-935-7
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Melting Point 40 - 45 °C
Molar Mass 563.76 g/mol
Flash Point 121 °C
pH  - 3 (50 g/l, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR