N-(2-Chlorobenzyloxycarbonyloxy)succinimide for synthesis, Sigma-Aldrich®

Warning

Synonyms: 2-Chlorobenzyl (2,5-dioxopyrrolidin-1-yl) carbonateN-(2-Chlorocarbobenzoxyoxy)succinimide2-Chlorobenzyl Succinimidyl CarbonateO-(2-Chlorobenzyloxycarbonyl)-N-hydroxysuccinimide

M-Clarity™ quality level = MQ200

8.40049.0010 8.40049.0050 8.40049.0010 8.40049.0050
EM8.40049.0010CS 7857.41 USD
EM8.40049.0010 CA8.40049.0050 CA8.40049.0010 EM8.40049.0050
N-(2-Chlorobenzyloxycarbonyloxy)succinimide for synthesis, Sigma-Aldrich®
N-(2-Chlorobenzyloxycarbonyloxy)succinimide
Formula: C₁₂H₁₀ClNO₅
MW: 283.66 g/mol
MDL Number: MFCD00010743
CAS Number: 65853-65-8

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 101 - 105 °C
Molar Mass 283.67 g/mol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR