Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.

2-Nitroimidazole for synthesis

Warning
Synonyms: 2-Nitro-1H-imidazole

8.24465.0500 8.24465.0500
EM8.24465.0500CS 229519.72 USD InStock
EM8.24465.0500 CA8.24465.0500
2-Nitroimidazole for synthesis
2-Nitroimidazole


Formula: C₃H₃N₃O₂
MW: 113,08 g/mol
Melting Pt: 279 - 284 °C
Storage Temperature: Ambient
MDL Number: MFCD00005185
CAS Number: 527-73-1
EINECS: 208-425-5
UN: 2811
ADR: 6.1,III
Merck Index: 13,00923
Order Now
Specification Test Results
Solubility (20 °C) slightly soluble
Melting Point 279 - 284 °C
Molar Mass 113.08 g/mol

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty