(+)-DIOP for synthesis, Sigma-Aldrich®

Warning

Synonyms: (((4S,5S)-2,2-Dimethyl-1,3-dioxolane-4,5-diyl)bis(methylene))bis(diphenylphosphine)(4S,5S)-(+)-4,5-Bis(diphenylphosphinomethyl)-2,2-dimethyl-1,3-dioxolane(S,S)-DIOP

M-Clarity™ quality level = MQ200

8.24429.0500 8.24429.0500
EM8.24429.0500CS 149002.24 USD
EM8.24429.0500 CA8.24429.0500
(+)-DIOP for synthesis, Sigma-Aldrich®
(+)-DIOP
Formula: C₃₁H₃₂O₂P₂
MW: 498.532 g/mol
MDL Number: MFCD00009760
CAS Number: 37002-48-5
EINECS: 253-307-9

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 86 - 89 °C
Molar Mass 498.54 g/mol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR