2-Vinylpyridine stabilized for synthesis, Sigma-Aldrich®

Danger


Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)
M-Clarity™ quality level = MQ200

8.22189.0250 8.22189.0005 8.22189.0250 8.22189.0005 8.22189.9025
EM8.22189.0250CS 1047.56 USD
CA8.22189.0005 CA8.22189.9025 EM8.22189.0250 CA8.22189.0250 EM8.22189.0005
2-Vinylpyridine stabilized for synthesis, Sigma-Aldrich®
2-Vinylpyridine
Formula: C₇H₇N
MW: 105.14 g/mol
Boiling Pt: 159 °C (1013 hPa)
Melting Pt: –50 °C
Density: 0.9735 g/cm³ (20 °C)
Flash Pt: 51 °C
Storage Temperature: Freezer
MDL Number: MFCD00006355
CAS Number: 100-69-6
EINECS: 202-879-8
UN: 3073
ADR: 6.1 (3, 8),II

Order Now

Specification Test Results

Solubility 27 g/l (20 °C)
Melting Point -50 °C
Molar Mass 105.14 g/mol
Boiling Point 158 - 160 °C (1013 hPa)
Vapor Pressure 2.5 hPa (20 °C)
Flash Point 51 °C
Refractive Index 1.5495 (20 °C, 589 nm)
Explosion Limit 1.3 - 10.7 %(V)
Density 0.97 g/cm3 (20 °C)
pH 8 (100 g/l, H2O, 20 °C)
Ignition Point 440 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR