1-Hexadecene for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.22064.0100 8.22064.0100 8.22064.0500 8.22064.0500
EM8.22064.0100CS 596 USD
EM8.22064.0100 CA8.22064.0500 CA8.22064.0100 EM8.22064.0500
1-Hexadecene for synthesis, Sigma-Aldrich®
1-Hexadecene
Formula: C₁₆H₃₂
MW: 224.43 g/mol
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: 4 °C
Density: 0.7811 g/cm³ (20 °C)
Flash Pt: 128 °C
Storage Temperature: Ambient
MDL Number: MFCD00008991
CAS Number: 629-73-2
EINECS: 211-105-8

Order Now

Specification Test Results

Solubility 0.0012 g/l (25 °C)
Melting Point 2.2 °C
Molar Mass 224.42 g/mol
Boiling Point 274 °C (1013 hPa)
Vapor Pressure 0.35 hPa (25 °C)
Flash Point 132 °C
Refractive Index 1.4401 (20 °C, 589 nm)
Explosion Limit 0.5 - 5.8 %(V)
Density 0.78 g/cm3 (20 °C)
Ignition Point 240 °C
Viscosity Kinematic 4.0 mm2/s (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR